missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Gr-1/Ly-6G, Fluorescein, Clone: RB6-8C5, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems FAB1037F-025
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.Specifications
| Gr-1/Ly-6G | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Ly6g | |
| The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
| 25 Tests | |
| Primary | |
| Detects mouse Gr-1/Ly-6G. Weak cross-reactivity with Ly-6C is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| RB6-8C5 | |
| Flow Cytometry 10 uL/10^6 cells | |
| C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58530 | |
| Mouse | |
| Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction