Learn More
anti-gamma H2AX (p Ser139), Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-384
Encompass
Product Code. 15764497
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
gamma H2AX Polyclonal specifically detects gamma H2AX in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP), Knockout Validated.