Learn More
Foxp3 Rat anti-Mouse, PE, Clone: MF23, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560408
Additional Details : Weight : 0.02000kg
Description
Foxp3 is a 50-55kDa protein also known as Forkhead box P3, JM2, or IPEX. It is a member of the forkhead or winged helix family of transcription factors and is specifically expressed by T regulatory (Treg) cells. Foxp3 has been reported to be a key regulatory protein for Treg cell development and function. Ectopic expression of Foxp3 in conventional T cells is sufficient to induce suppressive activity, repress the production of cytokines such as IL2 and IFN-γ, and upregulate Treg cell-associated molecules such as CD25, CTLA4 and GITR. It has been found that the mutation of Foxp3 is responsible for “scurfy” mice. When overexpressed, Foxp3 leads to poor T cell proliferation and activation.
Monoclonal antibody MF23 reacts with mouse Foxp3. Immunoblotting with MF23 antibody has confirmed it recognizes an epitope between 1-87 amino acids in the N-terminal domain.
Host Species: Rat
Clone: MF23
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse Foxp3 Recombinant Protein
Intracellular Staining
Specifications
Foxp3 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Foxp3 Recombinant Protein | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b |
Flow Cytometry | |
MF23 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.