missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Enolase 2/Neuron-specific Enolase, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-58870SS
This item is not returnable.
View return policy
Description
Enolase 2/Neuron-specific Enolase Polyclonal specifically detects Enolase 2/Neuron-specific Enolase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| Enolase 2/Neuron-specific Enolase | |
| Polyclonal | |
| Western Blot 1:2000, Simple Western 1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Knockdown Validated | |
| P09104 | |
| ENO2 | |
| Recombinant full length human NSE purified from E. coli. [UniProt# P09104] | |
| Unpurified | |
| RUO | |
| 2026 | |
| Human, Mouse, Rat | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Supplied as serum with 5mM Sodium Azide | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| Rabbit | |
| 47 kDa | |
| 0.025 mL | |
| Primary | |
| Mouse data from customer review. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only