Learn More
CD8 Mouse anti-Human, PE, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555367
Additional Details : Weight : 0.02000kg
Description
Clone RPA-T8 reacts with the α subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a are not cross-blocking. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
CD8 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human CD8a | |
100 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.