Learn More
CD49c Mouse, Unlabeled, Clone: 42, BD
Mouse Monoclonal Antibody
321.00€ - 504.00€
Specifications
Antigen | CD49c |
---|---|
Clone | 42 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15825779
|
BD Biosciences
611044 |
50 μg |
321.00€
1 set |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
15835779
|
BD Biosciences
611045 |
150 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Integrins are a family of dimeric proteins that mediate cell-to-cell and extracellular matrix adhesion. They consist of a large α chain that is non-covalently associated with a smaller β chain which defines the integrin subfamilies. VLA-3 (Very Late Antigen-3), a member of the integrin superfamily, exhibits elevated expression on B lymphocytes, but is also found on monocytes, platelets, and hematopoietic progenitor cells. A heterodimer of α3 (CD49c) and β1 (CD29) subunits, VLA is a receptor for laminin, fibronectin, and collagen. The α3 chain contains a large extracellular domain with three putative metal-binding sequences, a transmembrane domain, and a short cytoplasmic tail. Differing requirements for divalent cations and the influence of RGD peptides results in multiple ligand-binding mechanisms for VLA-3. Although its expression is restricted in normal tissues, VLA-3 is found on a variety of cultured tumor cells. In addition, levels of VLA-3 have been shown to correlate with the degree of invasiveness of malignant melanoma cells. Thus, VLA-3 mediates intercellular adhesion and cell migration in normal and, possibly, cancerous cell types.
Immunofluorescence, Western Blotting
Specifications
CD49c | |
250μg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Integrin α3; VLA-3α |
42 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Mouse, Rat | |
Mouse VLA-3α aa. 110-325 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.