missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36/SR-B3 Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 82 publications
Brand: Novus Biologicals NB400-144SS
This item is not returnable.
View return policy
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD36/SR-B3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Rabbit | |
| 75 kDa | |
| 0.025 mL | |
| Primary | |
| Human, bovine, mouse and monkey. Porcine reactivity reported in scientific literature (PMID: 23727393). Immunogen sequence has 95% identity to rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen | |
| P16671 | |
| CD36 | |
| A synthetic peptide mapping to a region of human CD36 between residues 100-200. [UniProt# P16671] | |
| Affinity Purified | |
| RUO | |
| 948 | |
| Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only