Learn More
CD21 Mouse anti-Human, APC, Clone: B-ly4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561357
Additional Details : Weight : 0.01000kg
Description
The B-ly4 monoclonal antibody specifically binds to CD21, a 145 kDa glycosylated type I integral membrane protein. CD21 is a receptor for the C3d complement fragment and for Epstein-Barr virus (EBV). CD21 is expressed on mature B cells, follicular dendritic cells, and some epithelial cells. It is also weakly expressed on the subset of mature T cells and thymocytes. CD21 plays a role in B-cell activation and proliferation. It may also play a role in modulating the function of T cells in the immune response to infections by lymphotropic viruses. Recently, CD21 was found to be part of a large complex containing CD19, CD81, and possibly other molecules.
Specifications
CD21 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Human, Porcine, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
B-ly4 | |
CR2; Complement receptor type 2; C3DR; EBV-R; Epstein-Barr virus receptor | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.