Learn More
anti-Cannabinoid R1/CB1/CNR1, PE, Clone: 368302, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB3834P
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.