Learn More
anti-Calreticulin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NB600-101SS
Description
Calreticulin Polyclonal specifically detects Calreticulin in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Electron Microscopy, Protein Array, Knockdown Validated, Block/Neutralize.Specifications
| Calreticulin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP | |
| Rabbit | |
| 48 kDa | |
| 0.025 mL | |
| Primary | |
| Human, bovine, rat and mouse calreticulin. Others not tested. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50 -1:200, Dot Blot, Electron Microscopy, Protein Array, Knockdown Validated, Block/Neutralize | |
| P14211 | |
| CALR | |
| Calreticulin Antibody was developed against a fusion protein to mouse Calreticulin [Uniprot: P14211] | |
| Affinity Purified | |
| RUO | |
| 811 | |
| Human, Mouse, Rat, Bovine, Hamster, Primate | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only