missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1q Rabbit anti-Human, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516601
Additional Details : Weight : 0.01000kg
Description
PA5-16601 targets C1q Complement in IF applications and shows reactivity with Human samples. The PA5-16601 immunogen is c1q component isolated from human serum.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.Specifications
C1q | |
Polyclonal | |
TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4 | |
P02745 | |
C1QA | |
C1q component isolated from human serum | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence | |
FITC | |
C1QA | |
complement C1q subcomponent subunit A, complement component 1, q subcomponent, alpha polypeptide, complement component C1q, A chain | |
Rabbit | |
Purified | |
RUO | |
712 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |