Learn More
STEMCELL Technologies Anti-Adherence Rinsing Solution
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Formerly known as AggreWell™ Rinsing Solution, name change effective Dec 2017
- Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion
- It can be used with plates, flasks, pipettes, strainers, and other cultureware
- Anti-Adherence Rinsing Solution is required for use with the following:
- AggreWell™ plates (e.g. Catalog #34811)
- Organoid growth media, including HepatiCult™ (Catalog #06030) and PancreaCult™ (Catalog #06040)
Specifications
Specifications
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.