missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Annexin V Apoptosis Kit, FITC, with Control Cells

Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)

Brand:  Novus Biologicals™ NBP2-29373-100Tests

Encompass

Product Code. 15706807

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Alert:

For Research Use Only

This item is not returnable. View return policy

Description

Description

  • Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells.
  • Annexin V staining, paired with 7-AAD or PI, is widely used to identify apoptotic stages by flow cytometry
Specifications

Specifications

Store at 4°C. Do not freeze.
FITC
Flow (Cell Surface), Immunocytochemistry/Immunofluorescence
100 Tests
Annexin V-FITC protein, Propidium Iodide (PI), 10X PBS, 10X Binding Buffer, Control Cells: 1x106 cells/mL
Apoptosis Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.