Learn More
Novus Biologicals™ Annexin V Apoptosis Kit [FITC]
Product Code. 18780284 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780284 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only
Product Code. 18780284 Supplier Novus Biologicals™ Supplier No. NBP229373100Tests
  • ONLINE EXCLUSIVE PRICE
    554.00€
    523.95€ / 100 tests
    Save 30.05€
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
For Research Use Only
Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
  • Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells.
  • Annexin V staining, paired with 7-AAD or PI, is widely used to identify apoptotic stages by flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. Do not freeze.
Quantity 100 Tests