missing translation for 'onlineSavingsMsg'
Learn More

Anisomycin, MP Biomedicals™

Product Code. 17640887 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 17640887

Brand: MP Biomedicals 0215988125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synonym: 2-[p-Methoxybenzyl]-3,4-Pyrrolidinediol 3-Acetate, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-Pyrrolidinediol-3-Acetate, Flagecidin

Anisomycin is an antiprotozoal agent that inhibits protein synthesis and activates a restricted subset of kinases currently only identified by p54 and MAP kinase. It can be used as a study aid for cytoplasmic signals resulting in nuclear signaling and induction of c-fos and c-jun. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22862-76-6

Specifications

Physical Form Powder
Source Streptomyces griseolus
Color White
Quantity 25 mg
Percent Purity ≥97%
compliance-icons
Product Identifier
  • Anisomycin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Anisomycin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.