Learn More
Anisole-d8, CDN
Product Code. 30316651
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316651
Brand: CDN CDND10241G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Anisole-d8
CAS 54887-54-6
Molecular Formula C7 D8 O
Quantity 1 g
Synonym 6-(Methoxy-d3)-benzene-1,2,3,4,5-d5, Methoxy-d3-benzene-d5, Perdeuteroanisole
InChI Formula InChI=1 S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D
SMILES [2 H]c1c([2 H])c([2 H])c(OC([2 H])([2 H])[2 H])c([2 H])c1[2 H]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethoxy)benzene
Molecular Weight (g/mol) 116.19
Formula Weight 116.1077 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity