Learn More
Laboratoires Anios™ Aniosgel 85 NPC
Click to view available options
Volume (Metric):
500 mL
1 L
Packaging:
Airless bottles
Pump Bottles
Unit Size:
Pack of 12
Description
- No allergenic substances, fragrance, or coloring
- Broad antimicrobial spectrum
- Hygienic treatment in 30 sec.
- For hygienic treatment and surgical disinfection
- Active against bacteria, mycobacteria, yeasts, molds, viruses
Tekniske data
Tekniske data
| Product Type | Sterilizer/Disinfectant Hand Gel |
| Certifications/Compliance (Europe) | EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791 |
| Composition | Ethanol in the presence of thickening, moisturizing and emollient agents, and water. |
| For Use With (Application) | Hygienic treatment and surgical disinfection of hands |
| Packaging | Airless bottles |
| Quantity | 12 Bottles |
| Volume (Metric) | 1 L |
Sikkerhed og håndtering
Produktidentifikator
- ANIOSGEL 85 NPC
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion