missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-like Protein 7/ANGPTL7 Mouse anti-Human, Clone: 121408, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB914-SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 7/ANGPTL7 Monoclonal specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Human samples. It is validated for Western Blot.Specifications
| Angiopoietin-like Protein 7/ANGPTL7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, ANGPTL7, AngX, CDT6, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 10218 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 121408 | |
| Western Blot 1 ug/mL | |
| O43827 | |
| ANGPTL7 | |
| E. coli-derived recombinant human Angiopoietin-like Protein 7/ANGPTL7 Lys28-Pro346 Accession # O43827 | |
| 25 μg | |
| Primary | |
| Detects human Angiopoietin-like Protein 7/ANGPTL7 in direct ELISAs and Western blots. In Western blots, less than 3% cross-reactivity with recombinant human Angiopoietin-1 is observed. | |
| Human | |
| Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction