missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Angiopep-2-Cys

Product Code. 30564816
Change view
Click to view available options
:
5 mg
Product Code.
30564816 5 mg
1 options
This item is not returnable. View return policy

Product Code. 30564816

Brand: Tocris Bioscience™ 8902/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Binds to LRP1 and facilitates receptor-mediated transcytosis of cargo across the BBB

Angiopep-2-Cys is a synthetic peptide able to specifically interact with low-density lipoprotein receptor-related protein 1 (LRP1) receptors present on brain endothelial cells and neurons. Angiopep-2-Cys is a conjugate of Angiopep-2 (Cat. No. 8828) and Cystein, and can be used to functionalize LNPs via thiol-maleimide click chemistry to target LRP1 receptors present on blood-brain barrier (BBB). mRNA LNPs functionalized with Angiopep-2-Cys shows improved transfection efficiency in cultured brain endothelial and neuronal cells, and in vivo, after systemic administration in mice.
TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
CAS 906480-09-9
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed: 2404.64
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.