Learn More
Fisherbrand™ Anemometer Compact
Shop All Fisherbrand Products
Click to view available options
Airflow Rate:
0.4 to 30 m/sec.
Unit Size:
Each
Specifications
Specifications
| Type | Compact Anemometer |
| Airflow Rate | 0.4 to 30 m/sec. |
| Case Material | ABS Plastic |
| Height (Metric) | 210 mm |
| Power Supply Details | 6V battery |
| Scale | m/s, ft/min, km/h, knots and mile/h |
| Temperature (Metric) | 0°C to 50°C |
| Weight (Metric) | 182 g |