Learn More
ABEBA™ Anatom Green/Yellow Safety Sandals S1P
missing translation for 'orderingAttributeHoverText'
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
missing translation for 'unitSize'
1 pair
Description
- Comply to EN ISO 20345: 2012, S1P, SRC
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable ACC wave insole
Specifications
Specifications
| Color | Green, Yellow |
| Material | Cordura |
| Size | 36 |
| Toe Material | Steel |
| Certifications/Compliance (Europe) | EN ISO 20345: 2012 S1P SRC |
| Gender | Men's |
| Ankle Style | Sandals |
| Waterproof | Water Resistant |
| Product Type | S1P SRC |