Learn More
ABEBA™ Anatom Black Safety Lace Up Shoes S3
Click to view available options
Size:
49
50
51
52
Unit Size:
1 pair
Description
- Comfort, robustness, durability
- Comply to EN ISO 20345 : 2012, S3, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU double density outsole
- Water resistant smooth black leather
- Steel toe cap (200 joules)
- Textile antipuncture midsole
- Scratch-resistant coating in toe and heel area
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- Removable ACC Wave insole
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2012 S3 SRC ESD |
| Closure Type | Laces |
| Color | Black |
| Eyelets Material | Stainless steel |
| Liner Material | Breathable silver ions lining |
| Material | Leather |
| Size | 50 |
| Sole Features | Slip and abrasion resistant, ESD dissipative, shock absorbent |
| Sole Material | Polyurethane2D |
| Style | Low-cut |
| Show More |