Learn More
Abeba™ Anatom 2615 Shoes
Product Code. 15668167
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15668167
Brand: Abeba™ 2615/46
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Microfiber, perforated
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Double velcro fastening
  • Reflective material
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Black
Material Microfiber
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2615
Toe Material Steel