Description
- Covers draft shield and console pad
- Suitable for a variety of EX, AX, PX, and PR model analytical balances
- An out-of-use cover
Specifications
Specifications
| For Use With (Equipment) | A variety of analytical balances including EX, AX, PX, and PR models |
| Color | Gray |
| Product Line | Dust cover |
| Type | Analytical Balance Cover |