Learn More
Amylose, TRC
Specifications
Specifications
| Chemical Name or Material | Amylose |
| CAS | 9005-82-7 |
| Molecular Formula | (C6 H10 O5)n |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16+,17-,18-/m1/s1 |
| SMILES | OC[C@H]1O[C@@H]([C@@H]([C@H]([C@@H]1O[C@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)CO)O)O)O[C@@H]3[C@H](O[C@@H]([C@@H]([C@H]3O)O)O)CO |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 504.4 |