missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Trihydrate, MP Biomedicals™

Product Code. p-7083049 Shop All MP Biomedical Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11474582

Brand: MP Biomedicals 0219014705

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Active as a broad-spectrum antibiotic. Ampicillin Trihydrate, MP Biomedicals is a semi-synthetic derivative of penicillin that is commonly used to select for ampicillin resistance in mutated and transformed cells. It is also used for enhancing luminol che

Ampicillin trihydrate is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure. It is a penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

  • Inactivated by beta-lactamases; therefore, a beta-lactamase inhibitor should be considered when using ampicillin
  • Antimicrobial spectrum: gram-negative and gram-positive bacteria.
    • Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin
    • Against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines
  • Bactericidal only to growing E. coli. Inhibits cell wall synthesis via cleavage of β-lactam ring of ampicillin by β-lactamase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7177-48-2
Molecular Formula C16H25N3O7S
Molecular Weight (g/mol) 403.45
InChI Key RXDALBZNGVATNY-CWLIKTDRSA-N
Synonym Aminobenzylpenicillin trihydrate, alpha-Aminobenzylpenicillin trihydrate ;D[-]-α-Aminobenzylpenicillin, 6-[D(-)-α-Aminophenylacetamido]penicillanic acid
PubChem CID 23565
ChEBI CHEBI:31209
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O

Specifications

Physical Form Powder
Melting Point 198°C to 200°C (literature)
Concentration Working Concentration: 100mg/liter for both gram positive and gram negative bacteria. It is typically stable in media at 37°C for approximately 3 days.
Quantity 5 g
Solubility Information Soluble in 1M Ammonium Hydroxide (50mg/mL), dilute acids (50mg/mL 1M HCl) or bases; slightly soluble in water (10mg/mL); practically insoluble in alcohol, chloroform, ether and fixed oils.
Formula Weight 403.46
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.