missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMN 082 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AMN 082 dihydrochloride is a the first selective mGlu7 agonist. Potently inhibits cAMP accumulation and stimulates GTPγS binding in recombinant cells and on membranes expressing mGlu7 (EC50 = 64 - 290 nM). Selective over other mGluR subtypes and selected ionotropic glutamate receptors up to 10 μM. Acts via a novel allosteric site and is orally active and brain penetrant. Reduces haloperidol-induced catalepsy in rats.
View important information regarding the usage of AMN 082 dihydrochloride .
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 465.45 |
| Purity | >99% |
| Molecular Weight (g/mol) | 465.462 |
| Chemical Name or Material | AMN 082 dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction