missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Ammonia Seawater Kit

Product Code. 11943727
Click to view available options
Range:
1 to 2mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11943727

Brand: Palintest™ AT170

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reagent required to prevent sea water interference with ammonia readings. Palintest™; Ammonia Seawater Kit uses salicylate (indophenol) chemistry.

  • Prevent sea water interference with ammonia readings.
  • Uses salicylate (indophenol) chemistry.

Specifications

For Use With (Application) Ammonia conditioning reagent
Range 1 to 2mg/L
Type Ammonium water sea kit
compliance-icons compliance-icons
Product Identifier
  • AMMONIA CONDITIONING REAGENT
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash contaminated skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see medical advice on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium tartrate dihydrate, lithium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.