Learn More
Ammonium Thiocyanate Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Product Code. 10254290 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10254290 2.5 L 2.5 L
10639672 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10254290 Supplier Fisher Chemical Supplier No. J/1500/PB17
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
MDL Number 11428
Specifications
Color Colorless
pH 4.5 to 6.0
Physical Form Liquid
Quantity 2.5 L
Packaging HDPE plastic bottle