Learn More
Thermo Scientific Chemicals Ammonium, Quant∣r Test Strips
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Fully miscible
- Liquid form
Specifications
Specifications
| Format | Liquid |
| For Use With (Application) | Reagents and strips to measure 10-400ppm NH4+1 in aqueous solution |
| Quantity | 1 Kit |
| Storage Requirements | Ambient |
| Type | Fully Miscible |