Learn More
Ammonium Persulfate, ACS, ≥98%, Ultrapure
Product Code. 15885228 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
10g
1kg
Product Code. 15885228
Brand: Thermo Scientific J76322.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High speed initiator used to catalyze acrylamide polymerization. | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
A high speed initiator used to catalyze acrylamide polymerization.
Working Stock Solution (10%):
  • Ammonium Persulfate - 1 gm
  • dH2O to 10 ml
  • This solution should be stored at 4°C and prepared fresh weekly.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
Specifications
Melting Point 120°C (decomposition)
Physical Form Crystalline Powder
Quantity 10 g
UN Number UN1444
Formula Weight 228.19
Percent Purity ≥98%
Grade ACS
Packaging Plastic bottle
Specific Gravity 1.982
Density 1.982
Chemical Name or Material Ammonium Persulfate