missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min

Product Code. 11326957
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326957 0.5 g 0.5g
11346957 1 g 1g
11336957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326957 Supplier Thermo Scientific Alfa Aesar Supplier No. 010823.02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

Ammonium hexachloroosmate(IV) is suitable for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-08-5
Molecular Formula Cl6H8N2Os
Molecular Weight (g/mol) 439.00
MDL Number MFCD00010883
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
PubChem CID 11729867
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 170°C (sublimation)
Color Brown to Black
Physical Form Powder
Quantity 0.5 g
Linear Formula (NH4)2OsCl6
Merck Index 14,536
Solubility Information Soluble in water.
IUPAC Name diazanium;hexachloroosmium(2-)
Formula Weight 438.99
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 42.8% min.
Sensitivity Hygroscopic
Density 2.9 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroosmate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.