Learn More
Ammonium Dihydrogen Orthophosphate, Technical, SLR, Fisher Chemical
Product Code. 10548990 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
Product Code. 10548990
Brand: Fisher Chemical A/4520/53
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Melting Point 190°C
CAS 7722-76-1
Color White
pH 3.8 to 4.4
Physical Form Solid
Quantity 500 g
Molecular Formula H6NO4P
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES N.OP(O)(O)=O
IUPAC Name phosphoric acid amine
Molecular Weight (g/mol) 115.03
Formula Weight 115.03g/mol
Packaging Plastic powder jar