Learn More
Alla France™ Ammonia Density Hydrometer
Click to view available options
Length (Metric):
300 mm
Unit Size:
Each
Description
Order Info
dimLengthMetric
Specifications
Specifications
| Quantity | 1Each |
| Type | Ammonia Density Hydrometer |
| Accuracy | ±0.004g/mL |
| Length (Metric) | 300 mm |
| Specific Gravity Range | 0.800 to 1.000 g/mL |