Learn More
WTW™ Ammonium Cell Test Kit
Product Code. 11771043
Click to view available options
Range:
0.010 - 2.000mg/I
Unit Size:
0.25kg
Product Code. 11771043
Brand: WTW™ 250495
With certificate for optimum precision.
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection
TRUSTED_SUSTAINABILITY
Specifications
Analytes Indophenol blue (Berthelot reaction)
For Use With (Application) For waste water, drinking water, sea water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
Includes Certificates
No. of Tests 25
Range 0.010 - 2.000mg/I
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Ammonium (NH4) cell test kit