Learn More
Novus Biologicals™ Amintra (R) Desalting Spin Columns
Shop All Bio Techne Products
Click to view available options
Quantity:
100 Units
50 Units
Unit Size:
50 units
Pack of 100
Specifications
Specifications
| Content And Storage | 4°C |
| Quantity | 50 Units |
| For Use With (Application) | Flow Cytometry |