missing translation for 'onlineSavingsMsg'
Learn More

Aminobutanol, TRC

Product Code. 30003775
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003775 10 g 10g
30138750 25 g 25g
30136271 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003775 Supplier Toronto Research Chemicals Supplier No. TRCA60211010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4 H11 N O
Molecular Weight (g/mol) 89.14
Synonym 1-Butanol, 2-amino-, 2-Amino-1-butanol, (RS)-2-Amino-1-butanol, (±)-2-Amino-1-butanol, 1-(Hydroxymethyl)propylamine, 1-Hydroxy-2-aminobutane, 1-Hydroxy-2-butylamine, 1-Hydroxy-sec-butylamine, 1-Hydroxybutan-2-amine, 2-Amino-1-hydroxybutane, 2-Aminobutyl alcohol, DL-2-Amino-1-butanol, DL-2-Aminobutanol, DL-α-Aminobutanol, NSC 1068, dl-2-Amino-1-butanol
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(N)CO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H11NO/c1-2-4(5)3-6/h4,6H,2-3,5H2,1H3
Formula Weight 89.0841
Chemical Name or Material 2-Amino-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.