missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde dimethyl acetal, 99%

Product Code. 11452887
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11452887

Brand: Thermo Scientific Alfa Aesar A15498.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22483-09-6 | C4H11NO2 | 105.137 g/mol

Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Solubility
Miscible in water. Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22483-09-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00008135
InChI Key QKWWDTYDYOFRJL-UHFFFAOYSA-N
Synonym aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
PubChem CID 89728
IUPAC Name 2,2-dimethoxyethanamine
SMILES COC(CN)OC

Specifications

Density 0.973
Boiling Point 135°C to 138°C
Flash Point 41°C (106°F)
Refractive Index 1.417
Quantity 100 g
UN Number UN2733
Beilstein 741868
Solubility Information Miscible in water. Soluble in chloroform,methanol.
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material Aminoacetaldehyde dimethyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoacetaldehyde dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aminoacetaldehyde dimethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.