Learn More
Cytiva Amersham™ Megaprime™ DNA Labeling System
Product Code. 10175804 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
30 reactions
60 reactions
For Use With (Application):
dCTP Labeling
dNTP Labeling
Unit Size:
30 reactions
60 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity For Use With (Application) Unit Size
10175804 60 reactions dCTP Labeling 60 reactions
10575155 60 reactions dNTP Labeling 60 reactions
10025174 30 reactions dNTP Labeling Each
10714367 30 reactions dCTP Labeling 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10175804 Supplier Cytiva Supplier No. RPN1607
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
For labeling DNA probes to specific activities >1 x 109dpm/μg in 10 minutes
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -15°C to -30°C
For Use With (Application) dCTP Labeling
Includes Primer solution 300μL, Labeling Buffered ATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 600μL, Nucleotide solutions dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA, Reaction Buffer A 10 x concentr
No. of Reactions 60
Product Type Megaprime DNA Labeling System, dCTP (for use with labeled dCTP)
Quantity 60 reactions