Learn More
Cytiva Amersham™ Hyperfilm™ MP
Product Code. 10218674 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
5 x 7 in.
18 x 24 cm
8 x 10 in.
Quantity:
100 Sheets/Box, Single film bag
50 Sheets/Box, Indiv. film envelope
50 Sheets/Box, Single film bag
Unit Size:
100 sheets
50 sheets
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Quantity Unit Size
10218674 18 x 24 cm 50 Sheets/Box, Single film bag 50 sheets
10627265 8 x 10 in. 100 Sheets/Box, Single film bag 100 sheets
10742837 8 x 10 in. 50 Sheets/Box, Single film bag 50 sheets
10034944 5 x 7 in. 50 Sheets/Box, Single film bag 50 sheets
10094984 18 x 24 cm 100 Sheets/Box, Single film bag 100 sheets
10084764 18 x 24 cm 50 Sheets/Box, Indiv. film envelope 50 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10218674 Supplier Cytiva Supplier No. 28906843
  • ONLINE EXCLUSIVE PRICE
    325.00€
    291.56€ / 50 sheets
    Save 33.44€
    10% Off
 Restricted Item
In Stock
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. Cytiva Life Sciences™ Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
  • High sensitivity for low concentration targets
  • Optimized for direct and indirect autoradiography with 3H, 14C, 32P, 33P, 35S, 125I
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images
  • High resolution at an economical price
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) 32P, 33P, 125I Southern, Northern, and Western blots. 32S, 33P, 35P sequencing. Fluorography of 3H, 14C, and 35S
Quantity 50 Sheets/Box, Single film bag
Dimensions (LxW) 18 x 24 cm