Learn More
Cytiva Amersham™ ECL™ Western Blotting Reagent Pack
Product Code. 10565335 Shop All Cytiva Products
Click to view available options
Quantity:
Each
Unit Size:
Each
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10565335
Brand: Cytiva RPN2124
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Includes
anti-mouse IgG horseradish peroxidase conjugate, 100μL; anti-rabbit IgG horseradish peroxidase conjugate, 100μL; Amersham ECL blocking agent, 5g
Reagents for membrane blocking and secondary detection of membrane-bound mouse and rabbit primary antibodies
  • Designed for use with nitrocellulose or PVDF membranes
  • System optimized for use with ECL detection reagents (Not included)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) High sensitivity western blotting and chemifluorescent detection on scanners
Includes 100μL Anti-Mouse IgG, Peroxidase linked whole Antibody (from Sheep), 100μL Anti-Rabbit IgG, Peroxidase linked whole Antibody (from Donkey), 5g ECL™ membrane blocking agent
Quantity Each