Learn More
Cytiva Amersham™ AlkPhos Direct™ ECF Detection Module
Product Code. 10208724 Shop All Cytiva Products
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. 10208724
Brand: Cytiva RPN3685
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star™ or ECF™ detection can be used. DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.
  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • ECF detection for quantitation experiments
  • Rapid protocol saves two to three hours over conventional indirect methods
Order Info
Each system include the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500cm2: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, ECF detection reagents.
TRUSTED_SUSTAINABILITY
Specifications
Sufficient For 2500cm2 membrane
Includes Sufficient for 25 or 50 labeling reactions and detection of 2500cm2 membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, ECF detection reagents.
Quantity Each
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2