Learn More
Amberlite™ IRA-200C(Na), ion exchange resin
Product Code. p-4412892
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
Product Code. 11379907
Brand: Thermo Scientific Alfa Aesar 042252.30
Request Bulk or Custom Format
Styrene-DVB; Strongly acidic macroreticular cation exchange resin | Styrene-DVB
Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Amberlite IRA-200C(Na)
Physical Form Beads
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD01864001
Color Amber
Quantity 250 g
Solubility Information Insoluble in water.