Learn More
AmberChrom™, 50WX2, 200-400 mesh, H-form, Thermo Scientific™
Product Code. 17505625 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505625 100 g 100g
17515625 500 g 500g
17595615 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17505625 Supplier Thermo Scientific Acros Supplier No. 473031000
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Chemical Identifiers
CAS 69011-20-7 , 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *
Specifications
Physical Form Beads
CAS Min % 18%, 74%
CAS Max % 26%, 74%
Ion Exchange Capacity ≥0.6 eq/L (H form)
Particle Size 200 to 400 mesh, ≤15 % (< 50 μm), ≤15 % (> 210 μm)
Packaging Plastic Bottle
Water Retention Capacity 74 to 82 % (H form)
Color White to Yellow to Orange
Quantity 100 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™