Learn More
Emma Safety Footwear Amber Safety Shoes
Product Code. 15949101
Click to view available options
Size:
35
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 15949101
Brand: Emma Safety Footwear MM94251638
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
  • S1P-Safety shoe with sole plate for dry work
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD: Electrostatic Discharge
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Amber
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 38
Toe Material Steel