Learn More
Buerkle™ Aluminum Support Stand for Buerkle™ Jerricans
Product Code. 10334022
Click to view available options
For Use With (Equipment):
Support Stand for 2 Compact Jerrycans 5 L
Support Stand for 3 Compact Jerrycans 5 L
Unit Size:
Each
Product Code. 10334022
Brand: Buerkle™ 04359102
Only null left
Add to basket
Add to basket
Safe for storage
  • Support stand
  • For Compact Jerry Can 5L
  • For safe storage
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Support Stand for 2 Compact Jerrycans 5 L
For Use With (Equipment) Support Stand for 2 Compact Jerrycans 5 L
Material Aluminum
Product Type Jerry Can Support Stand