Learn More
Hellma™ Aluminum Spacer
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Specifications
Specifications
| Type | Spacer |
| Material | Aluminum |
| Quantity | 1 Ea. |
| Height (Metric) | 9 mm |
| Width (Metric) | 12.5 mm |