Learn More
Korff™ Aluminum Small Rolls
Product Code. p-8008778
Click to view available options
Width (Metric):
300mm
450mm
Description:
Foil Gauge: 0.015mm
Foil Gauge: 0.030mm
Unit Size:
10m
Product Code. 10523282
Brand: Korff™ 60010