Learn More
VACUUBRAND™ Aluminum Reducing Cross Piece With Small KF Flanges
Click to view available options
Diameter (Metric) Inside:
25/25/16/16 mm
40/40/16/16 mm
Size:
35 x 70 x 35 x 70 mm
45 x 90 x 65 x 130 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inside | 40/40/16/16 mm |
| Material | Aluminum |
| Quantity | 1 Each |
| Size | 45 x 90 x 65 x 130 mm |
| Type | Reducing Cross Piece |