Learn More
Brady™ Aluminum with Polyester Overlaminate Mandatory Signs
Product Code. 11764894
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Displayed Text:
Protective Gloves
Use Head Protection
Use breathing protection
Use face shield
Use full protective
Use overshoes
Use protective hood
Using Foot
Wear protective apron
Unit Size:
Each
Product Code. 11764894
Brand: Brady™ 257923
Only null left
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC 267-DIA 100-B7525
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
Diameter (Metric) 100 mm
For Use With (Application) Indoor and outdoor safety signs
Thickness 0.271 mm
Temperature Range (Metric) -40°C to 140°C
Shape Round
Displayed Text Use breathing protection
Product Type Mandatory Sign
Color White on Blue